accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NHONNHClHCl

CAS RN 1347744-96-0

IUPAC Name: N-(3-methylphenyl)-2-[4-(pyridin-2-yl)piperidin-1-yl]acetamide dihydrochloride
Synonyms: N-(3-methylphenyl)-2-[4-(pyridin-2-yl)piperidin-1-yl]acetamide dihydrochloride
Molecular Weight (g/mol): 382.33
Molecular Formula: C19H25Cl2N3O
InChi Key: KIGNSSHMCNBHQX-UHFFFAOYSA-N
SMILES: Cl.Cl.CC1=CC(NC(=O)CN2CCC(CC2)C2=CC=CC=N2)=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)