accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OHN

CAS RN 134865-74-0

IUPAC Name: N-(4-phenyl-1,2,3,4-tetrahydronaphthalen-2-yl)propanamide
Synonyms: N-(4-phenyl-1,2,3,4-tetrahydronaphthalen-2-yl)propanamide
Molecular Weight (g/mol): 279.38
Molecular Formula: C19H21NO
InChi Key: RCYLUNPFECYGDW-UHFFFAOYNA-N
SMILES: CCC(=O)NC1CC(C2=CC=CC=C2)C2=CC=CC=C2C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)