Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ 4-P-PDOT
GREENER_CHOICESDP

Catalog No. 103450 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
10-345-0 50 mg
10-341-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 10-345-0 Supplier R&D Systems™ Supplier No. 1034/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. MT2 antagonist

Melatonin receptor antagonist, >300-fold selective for the MT2 (vs MT1) subtype.

Chemical Identifiers

CAS 134865-74-0
Molecular Formula C19H21NO

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 279.38
Purity >99%
Molecular Weight (g/mol) 279.383
Chemical Name or Material 4-P-PDOT
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.