accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1354039-86-3

IUPAC Name: 2-{1-[(3,4-dimethoxyphenyl)methyl]-6,7-dimethoxy-1,2,3,4-tetrahydroisoquinolin-2-yl}-2-phenyl-N-(propan-2-yl)acetamide
Synonyms: 2-{1-[(3,4-dimethoxyphenyl)methyl]-6,7-dimethoxy-3,4-dihydro-1H-isoquinolin-2-yl}-N-isopropyl-2-phenylacetamide
Molecular Weight (g/mol): 518.65
Molecular Formula: C31H38N2O5
InChi Key: HXHOBPVRRPCTLG-UHFFFAOYNA-N
SMILES: COC1=C(OC)C=C(CC2N(CCC3=CC(OC)=C(OC)C=C23)C(C(=O)NC(C)C)C2=CC=CC=C2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results