accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CONHOHOHNNNF

CAS RN 1354235-96-3

IUPAC Name: 1-(3-{[4-(2-fluorophenyl)piperazin-1-yl]methyl}-4-methoxyphenyl)-1H,2H,3H,4H,9H-pyrido[3,4-b]indole-3-carboxylic acid
Synonyms: 1-(3-{[4-(2-fluorophenyl)piperazin-1-yl]methyl}-4-methoxyphenyl)-1H,2H,3H,4H,9H-pyrido[3,4-b]indole-3-carboxylic acid
Molecular Weight (g/mol): 514.6
Molecular Formula: C30H31FN4O3
InChi Key: FUHCEERDBRGPQZ-UHFFFAOYNA-N
SMILES: COC1=C(CN2CCN(CC2)C2=CC=CC=C2F)C=C(C=C1)C1NC(CC2=C1NC1=CC=CC=C21)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)