accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClClOONFCl

CAS RN 1355026-60-6

IUPAC Name: 2-chloro-3-[2-(2,4-dichlorophenoxy)ethoxy]-6-(fluoromethyl)pyridine
Synonyms: 2-chloro-3-[2-(2,4-dichlorophenoxy)ethoxy]-6-(fluoromethyl)pyridine
Molecular Weight (g/mol): 350.6
Molecular Formula: C14H11Cl3FNO2
InChi Key: FHPOTBQOUBMMCI-UHFFFAOYSA-N
SMILES: FCC1=NC(Cl)=C(OCCOC2=C(Cl)C=C(Cl)C=C2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)