Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Description
Potent sphingosine-1-phosphate receptor 4 (S1P4) agonist (EC50 = 45 nM). Displays no activity at other S1P receptor subtypes (S1P1-3 and S1P5) at concentrations up to 25 μM.
Specifications
Specifications
| Quantity | 50 mg |
| Formula Weight | 350.6 |
| Purity | >98% |
| Molecular Weight (g/mol) | 350.595 |
| Chemical Name or Material | CYM 50260 |
Safety and Handling
| Recommended Storage | Store at -20°C |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?

