accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNNONONHNNOCH3

CAS RN 1355244-02-8

IUPAC Name: N-{3-[4-(4-methoxyphenyl)piperazin-1-yl]propyl}-2-{13-methyl-7-oxo-2,8,10-triazatricyclo[7.4.0.0²,⁶]trideca-1(13),3,5,9,11-pentaen-8-yl}acetamide
Synonyms: N-{3-[4-(4-methoxyphenyl)piperazin-1-yl]propyl}-2-{13-methyl-7-oxo-2,8,10-triazatricyclo[7.4.0.0²,⁶]trideca-1(13),3,5,9,11-pentaen-8-yl}acetamide
Molecular Weight (g/mol): 488.59
Molecular Formula: C27H32N6O3
InChi Key: PMTIWRPLQBVEMR-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1)N1CCN(CCCNC(=O)CN2C(=O)C3=CC=CN3C3=C(C)C=CN=C23)CC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results