CAS RN 1358575-02-6
CAS RN 1358575-02-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Tocris Bioscience™ FTBMT

Potent and selective GPR52 agonist; orally bioavailable and BBB permeable