accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NONH3CNNHOH2C

CAS RN 136199-02-5

IUPAC Name: 1,3-dipropyl-8-{tricyclo[3.3.1.0³,⁷]nonan-3-yl}-2,3,6,7-tetrahydro-1H-purine-2,6-dione
Synonyms: rolofylline
Molecular Weight (g/mol): 356.47
Molecular Formula: C20H28N4O2
InChi Key: PJBFVWGQFLYWCB-UHFFFAOYNA-N
SMILES: CCCN1C2=C(NC(=N2)C23CC4CC2CC(C3)C4)C(=O)N(CCC)C1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results