accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CONOHCl

CAS RN 136647-02-4

IUPAC Name: 4-[(diphenylmethoxy)methyl]-1-[3-(4-methoxyphenyl)propyl]piperidine hydrochloride
Synonyms: 4-[(diphenylmethoxy)methyl]-1-[3-(4-methoxyphenyl)propyl]piperidine hydrochloride
Molecular Weight (g/mol): 466.06
Molecular Formula: C29H36ClNO2
InChi Key: ZZSQARULZYQMIG-UHFFFAOYSA-N
SMILES: Cl.COC1=CC=C(CCCN2CCC(COC(C3=CC=CC=C3)C3=CC=CC=C3)CC2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results