accessibility menu, dialog, popup

UserName

Viewing profile as user:
NNNNHNOOH

CAS RN 1373422-53-7

IUPAC Name: 3-{[2-(pyridin-2-yl)-6-(2,3,4,5-tetrahydro-1H-3-benzazepin-3-yl)pyrimidin-4-yl]amino}propanoic acid
Synonyms: 3-{[2-(pyridin-2-yl)-6-(1,2,4,5-tetrahydro-3-benzazepin-3-yl)pyrimidin-4-yl]amino}propanoic acid
Molecular Weight (g/mol): 389.46
Molecular Formula: C22H23N5O2
InChi Key: AVZCPICCWKMZDT-UHFFFAOYSA-N
SMILES: OC(=O)CCNC1=NC(=NC(=C1)N1CCC2=CC=CC=C2CC1)C1=CC=CC=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)