Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ GSK J1
GREENER_CHOICESDP

Catalog No. 459350 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45-935-0 50 mg
45-931-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 45-935-0 Supplier R&D Systems™ Supplier No. 4593/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Potent JMJD3/UTX inhibitor

Potent inhibitor of the H3K27 histone demethylases JMJD3 (KDM6B) and UTX (KDM6A) (IC50 values are 28 and 53 nM respectively). Also inhibits KDM5B, KDM5C and KDM5A (IC50 values are 170, 550 and 6,800 nM respectively). Exhibits no activity against a panel of other histone demethylases (IC50 >20 μM), and displays no significant inhibitory activity against 100 protein kinases at a concentration of 30 μM.

Ethyl ester derivative and Negative Control also available.

Chemical Identifiers

CAS 1373422-53-7
Molecular Formula C22H23N5O2

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 389.45
Purity >99%
Molecular Weight (g/mol) 389.459
Chemical Name or Material GSK J1
Recommended Storage Desiccate at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.