CAS RN 1374356-45-2
IUPAC Name:
3-[1-(2,6-dichloro-3-fluorophenyl)ethoxy]-5-[1-(piperidin-4-yl)-1H-pyrazol-4-yl]pyridin-2-amine
Synonyms:
crizotinib
Molecular Weight (g/mol):
450.34
Molecular Formula:
C21H22Cl2FN5O
InChi Key:
KTEIFNKAUNYNJU-UHFFFAOYNA-N
SMILES:
CC(OC1=C(N)N=CC(=C1)C1=CN(N=C1)C1CCNCC1)C1=C(Cl)C=CC(F)=C1Cl
1
–
1
of
1
results
1
R&D Systems™ (S)-Crizotinib, >98%
CAS: 1374356-45-2 Molecular Formula: C21H22Cl2FN5O