Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ (S)-Crizotinib, >98%
GREENER_CHOICESDP

Catalog No. 602550 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-255-0 50 mg
60-251-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 60-255-0 Supplier R&D Systems™ Supplier No. 6025/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. MTH1 inhibitor

Potent MTH1 inhibitor (IC50 = 72 nM). Induces DNA damage and disrupts nucleotide pool homeostasis in cancer cells. Also attenuates colony formation of KRAS-mutated PANC1 cells in vitro. Suppresses tumor growth ∽50% in a colon cancer carcinoma xenograft model.

Chemical Identifiers

CAS 1374356-45-2
Molecular Formula C21H22Cl2FN5O

Specifications

Quantity 50 mg
Solubility Soluble to 50mM in DMSO and to 20mM in 1eq. HCl with gentle warming
Product Line Tocris
Formula Weight 450.34
Purity >98%
Molecular Weight (g/mol) 450.339
Chemical Name or Material 3-[(1S)-1-(2,6-Dichloro-3-fluorophenyl)ethoxy]-5-[1-(4-piperidinyl)-1H-pyrazol-4-yl]-2-pyridinamine
Recommended Storage Store at RT
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.