accessibility menu, dialog, popup

UserName

Viewing profile as user:
NOOOOOHOOFFFFFF

CAS RN 1380424-42-9

IUPAC Name: 1,1,1,3,3,3-hexafluoropropan-2-yl 4-[bis(2H-1,3-benzodioxol-5-yl)(hydroxy)methyl]piperidine-1-carboxylate
Synonyms: 1,1,1,3,3,3-hexafluoropropan-2-yl 4-[bis(2H-1,3-benzodioxol-5-yl)(hydroxy)methyl]piperidine-1-carboxylate
Molecular Weight (g/mol): 549.42
Molecular Formula: C24H21F6NO7
InChi Key: SXHQLPHDBLTFPM-UHFFFAOYSA-N
SMILES: OC(C1CCN(CC1)C(=O)OC(C(F)(F)F)C(F)(F)F)(C1=CC2=C(OCO2)C=C1)C1=CC2=C(OCO2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)