Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ KML 29
GREENER_CHOICESDP

Catalog No. 487250 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
48-725-0 50 mg
48-721-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 48-725-0 Supplier R&D Systems™ Supplier No. 4872/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Highly potent and selective MAGL inhibitor

Highly selective and potent monoacylglycerol lipase (MAGL) inhibitor. Exhibits potent inhibition of human, mouse and rat MAGL (IC50 values are 5.9, 15 and 43 nM, respectively). Exhibits no detectable inhibition of FAAH (IC50 > 50000 nM). Potently and selectively blocks hydrolysis of 2-arachidonoylglycerol (2-AG) in mice (IC50 = 2.5 nM and >50 μM for 2-AG and AEA respectively).

Chemical Identifiers

CAS 1380424-42-9
Molecular Formula C24H21F6NO7

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 549.42
Molecular Weight (g/mol) 549.422
Chemical Name or Material KML 29
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.