CAS RN 138847-85-5
IUPAC Name:
2-(morpholin-4-yl)ethyl 1-phenylcyclohexane-1-carboxylate
Synonyms:
2-(morpholin-4-yl)ethyl 1-phenylcyclohexane-1-carboxylate
Molecular Weight (g/mol):
317.43
Molecular Formula:
C19H27NO3
InChi Key:
RQHKZUBCUZVZEF-UHFFFAOYSA-N
SMILES:
O=C(OCCN1CCOCC1)C1(CCCCC1)C1=CC=CC=C1
1
–
1
of
1
results
1
R&D Systems™ PRE-084 hydrochloride
CAS: 138847-85-5 Molecular Formula: C19H27NO3