CAS RN 1391052-28-0
IUPAC Name:
3-[(3-chlorophenoxy)methyl]-1-(oxan-4-yl)-1H-pyrazolo[3,4-d]pyrimidin-4-amine hydrochloride
Synonyms:
3-(3-chlorophenoxymethyl)-1-(oxan-4-yl)pyrazolo[3,4-d]pyrimidin-4-amine hydrochloride
Molecular Weight (g/mol):
396.27
Molecular Formula:
C17H19Cl2N5O2
InChi Key:
QZXZQMUZEHTFHD-UHFFFAOYSA-N
SMILES:
Cl.NC1=C2C(COC3=CC(Cl)=CC=C3)=NN(C3CCOCC3)C2=NC=N1
1
–
1
of
1
results
1
R&D Systems™ PF 4800567 hydrochloride
CAS: 1391052-28-0 Molecular Formula: C17H19Cl2N5O2