accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1392399-03-9

IUPAC Name: N-(1-benzylpiperidin-4-yl)-6,7-dimethoxy-2-(4-methyl-1,4-diazepan-1-yl)quinazolin-4-amine trihydrochloride
Synonyms: N-(1-benzylpiperidin-4-yl)-6,7-dimethoxy-2-(4-methyl-1,4-diazepan-1-yl)quinazolin-4-amine trihydrochloride
Molecular Weight (g/mol): 600.03
Molecular Formula: C28H41Cl3N6O2
InChi Key: FMURUEPQXKJIPS-UHFFFAOYSA-N
SMILES: Cl.Cl.Cl.COC1=CC2=NC(=NC(NC3CCN(CC4=CC=CC=C4)CC3)=C2C=C1OC)N1CCCN(C)CC1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results