accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 140-41-0

H3CNCH3ONHClHOOClClCl

CAS RN 140-41-0

IUPAC Name: 1-(4-chlorophenyl)-3,3-dimethylurea; trichloroacetic acid
Synonyms: monuron-TCA
Molecular Weight (g/mol): 362.03
Molecular Formula: C11H12Cl4N2O3
InChi Key: DUQGREMIROGTTD-UHFFFAOYSA-N
SMILES: OC(=O)C(Cl)(Cl)Cl.CN(C)C(=O)NC1=CC=C(Cl)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results