accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 140158-50-5

IUPAC Name: 2-amino-3-(5-tert-butyl-3-oxo-2,3-dihydro-1,2-oxazol-4-yl)propanoic acid
Synonyms: 2-amino-3-(5-tert-butyl-3-oxo-2H-1,2-oxazol-4-yl)propanoic acid
Molecular Weight (g/mol): 228.25
Molecular Formula: C10H16N2O4
InChi Key: PIXJURSCCVBKRF-UHFFFAOYNA-N
SMILES: CC(C)(C)C1=C(CC(N)C(O)=O)C(=O)NO1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results