CAS RN 140187-25-3
IUPAC Name:
2-amino-3-(5-iodo-2,4-dioxo-1,2,3,4-tetrahydropyrimidin-1-yl)propanoic acid
Synonyms:
iodo-willardiine
Molecular Weight (g/mol):
325.06
Molecular Formula:
C7H8IN3O4
InChi Key:
AXXYLTBQIQBTES-UHFFFAOYNA-N
SMILES:
NC(CN1C=C(I)C(=O)NC1=O)C(O)=O
1
–
1
of
1
results
1
R&D Systems™ (S)-(-)-5-Iodowillardiine
CAS: 140187-25-3 Molecular Formula: C7H8IN3O4