Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ (S)-(-)-5-Iodowillardiine
GREENER_CHOICESDP

Catalog No. 030750 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
03-075-0 50 mg
03-071-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 03-075-0 Supplier R&D Systems™ Supplier No. 0307/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Highly potent and subtype-selective kainate agonist

Demonstrates high affinity for the kainate receptor subtype hGluK1 (formerly hGluR5) (Ki = 0.24 nM) and 600-4000-fold selectivity over both the AMPA receptor subtypes and the homomeric kainate receptor hGluK2 (formerly hGluR6).

Please refer to IUPHAR Guide to Pharmacology for the most recent naming conventions.

Chemical Identifiers

CAS 140187-25-3
Molecular Formula C7H8IN3O4

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 325.06
Purity >98%
Molecular Weight (g/mol) 325.062
Chemical Name or Material (S)-(-)-5-Iodowillardiine
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.