accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNNHSOOOH3CHNO

CAS RN 1403764-72-6

IUPAC Name: 2-methoxy-N-(3-methyl-2-oxo-1,2,3,4-tetrahydroquinazolin-6-yl)benzene-1-sulfonamide
Synonyms: 2-methoxy-N-(3-methyl-2-oxo-1,4-dihydroquinazolin-6-yl)benzenesulfonamide
Molecular Weight (g/mol): 347.39
Molecular Formula: C16H17N3O4S
InChi Key: TXZPMHLMPKIUGK-UHFFFAOYSA-N
SMILES: COC1=CC=CC=C1S(=O)(=O)NC1=CC2=C(NC(=O)N(C)C2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results