accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OO(S)NHO(S)NHO(R)HNO(R)NHO(S)NHO(R)HNO(R)H2NOHOOHCH3(R)HOOO(R)O(S)(R)NH2(R)HO(S)H3COO(S)O(R)OO(S)CH3(R)HO(R)HO(R)HO(S)HO(S)HO(R)O(S)O(S)HO(R)HO(R)OH(R)O(S)O(R)OH(R)HO(S)HOOHO(S)O(R)OH(S)OH(S)HO(S)HOOH(?)

CAS RN 1404-55-3

IUPAC Name: methyl (1S,2R,18R,19R,22R,34S,37R,40R,52S)-22-amino-2-{[(2S,4R,5R,6S)-4-amino-5-hydroxy-6-methyloxan-2-yl]oxy}-64-{[(2S,3R,4S,5S,6R)-3-{[(2S,3R,4R,5R,6S)-4,5-dihydroxy-6-(hydroxymethyl)-3-{[(2S,3S,4R,5R)-3,4,5-trihydroxyoxan-2-yl]oxy}oxan-2-yl]oxy}-4,5-dihydroxy-6-({[(3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyloxan-2-yl]oxy}methyl)oxan-2-yl]oxy}-18,26,31,44,49-pentahydroxy-30-methyl-21,35,38,54,56,59-hexaoxo-47-{[(2S,3S,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}-7,13,28-trioxa-20,36,39,53,55,58-hexaazaundecacyclo[38.14.2.2³,⁶.2¹⁴,¹⁷.2¹⁹,³⁴.1⁸,¹².1²³,²⁷.1²⁹,³³.1⁴¹,⁴⁵.0¹⁰,³⁷.0⁴⁶,⁵¹]hexahexaconta-3,5,8(64),9,11,14,16,23,25,27(61),29(60),30,32,41,43,45(57),46,48,50,62,65-henicosaene-52-carboxylate
Synonyms: ristocetin
Molecular Weight (g/mol): 2067.94
Molecular Formula: C95H110N8O44
InChi Key: VUOPFFVAZTUEGW-FBMDODLCSA-N
SMILES: COC(=O)[C@H]1NC(=O)[C@H]2NC(=O)[C@H](NC(=O)[C@@H]3NC(=O)[C@H]4NC(=O)[C@H](NC(=O)[C@H](N)C5=CC=C(O)C(OC6=CC4=CC(O)=C6C)=C5)[C@H](O)C4=CC=C(OC5=CC3=CC(OC3=CC=C(C=C3)[C@H]2O[C@@H]2C[C@@H](N)[C@@H](O)[C@H](C)O2)=C5O[C@@H]2O[C@H](COC3O[C@@H](C)[C@H](O)[C@@H](O)[C@H]3O)[C@@H](O)[C@H](O)[C@H]2O[C@@H]2O[C@@H](CO)[C@H](O)[C@@H](O)[C@H]2O[C@@H]2OC[C@@H](O)[C@@H](O)[C@@H]2O)C=C4)C2=CC=C(O)C(=C2)C2=C(O[C@@H]3O[C@H](CO)[C@@H](O)[C@H](O)[C@@H]3O)C=C(O)C=C12
Color 
  • (1)
Grade 
  • (1)
Percent Purity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results