accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 141993-16-0

H3CON(R)(S)(R)SOOH3CH3C

CAS RN 141993-16-0

IUPAC Name: (1R,5R,7S)-4-acetyl-10,10-dimethyl-3λ⁶-thia-4-azatricyclo[5.2.1.0¹,⁵]decane-3,3-dione
Synonyms: (1R,5R,7S)-4-acetyl-10,10-dimethyl-3λ⁶-thia-4-azatricyclo[5.2.1.0¹,⁵]decane-3,3-dione
Molecular Weight (g/mol): 257.35
Molecular Formula: C12H19NO3S
InChi Key: FECARJHVTMSYSJ-UMNHJUIQSA-N
SMILES: CC(=O)N1[C@@H]2C[C@@H]3CC[C@@]2(CS1(=O)=O)C3(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results