accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1421365-63-0

IUPAC Name: 5-chloro-2-(9H-fluoren-9-yl)-4-(4-methoxyphenoxy)-2,3-dihydropyridazin-3-one
Synonyms: 5-chloro-2-(9H-fluoren-9-yl)-4-(4-methoxyphenoxy)pyridazin-3-one
Molecular Weight (g/mol): 416.86
Molecular Formula: C24H17ClN2O3
InChi Key: QHVSQUYCVUHYKT-UHFFFAOYSA-N
SMILES: COC1=CC=C(OC2=C(Cl)C=NN(C3C4=CC=CC=C4C4=CC=CC=C34)C2=O)C=C1
Color 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results