accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2NONN(S)(S)HO(R)HOCH2HN

CAS RN 142217-69-4

IUPAC Name:
Synonyms:
Molecular Weight (g/mol):
Molecular Formula:
InChi Key:
SMILES:
Product Type 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)