accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 142326-59-8

IUPAC Name: 7-chloro-2-hydroxy-3-(3-phenoxyphenyl)-1,4-dihydroquinolin-4-one
Synonyms: 7-chloro-2-hydroxy-3-(3-phenoxyphenyl)-1H-quinolin-4-one
Molecular Weight (g/mol): 363.8
Molecular Formula: C21H14ClNO3
InChi Key: FLVRDMUHUXVRET-UHFFFAOYSA-N
SMILES: OC1=C(C(=O)C2=CC=C(Cl)C=C2N1)C1=CC=CC(OC2=CC=CC=C2)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)