accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOHPOHOHOPOHHOO

CAS RN 142523-38-4

IUPAC Name: [1-(4-hydroxyphenoxy)-1-phosphonoethyl]phosphonic acid
Synonyms: 1-(4-hydroxyphenoxy)-1-phosphonoethylphosphonic acid
Molecular Weight (g/mol): 298.12
Molecular Formula: C8H12O8P2
InChi Key: JKOCAAWWDVHWKB-UHFFFAOYSA-N
SMILES: CC(OC1=CC=C(O)C=C1)(P(O)(O)=O)P(O)(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results