CAS RN 1431148-26-3
CAS RN 1431148-26-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Tocris Bioscience™ Cyanine 5 Tyramide
Red fluorescent reagent widely used for signal amplification in IHC and FISH