CAS RN 1432043-31-6
IUPAC Name:
4-[(3,5-dihydroxyphenyl)methyl]-N-[(2-methyl-4-{4-methyl-2,4,5,9-tetraazatricyclo[8.4.0.0³,⁷]tetradeca-1(14),2,6,10,12-pentaene-9-carbonyl}phenyl)methyl]piperazine-1-carboxamide dihydrochloride
Synonyms:
4-[(3,5-dihydroxyphenyl)methyl]-N-[(2-methyl-4-{4-methyl-2,4,5,9-tetraazatricyclo[8.4.0.0³,⁷]tetradeca-1(14),2,6,10,12-pentaene-9-carbonyl}phenyl)methyl]piperazine-1-carboxamide dihydrochloride
Molecular Weight (g/mol):
654.59
Molecular Formula:
C32H37Cl2N7O4
InChi Key:
OFWMUAWQQXSNJI-UHFFFAOYSA-N
SMILES:
Cl.Cl.CN1NC=C2CN(C(=O)C3=CC(C)=C(CNC(=O)N4CCN(CC5=CC(O)=CC(O)=C5)CC4)C=C3)C3=CC=CC=C3N=C12
1
–
1
of
1
results
1
R&D Systems™ WAY 267464 dihydrochloride
CAS: 1432043-31-6 Molecular Formula: C32H37Cl2N7O4