Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ WAY 267464 dihydrochloride
GREENER_CHOICESDP

Catalog No. 393310 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
39-331-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 39-331-0 Supplier R&D Systems™ Supplier No. 3933/10

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Potent non-peptide oxytocin receptor agonist

Potent, selective non-peptide oxytocin receptor (OTR) agonist (Ki = 58.4 nM at human OTR). Exhibits anxiolytic-like effects. May have therapeutic potential for the treatment of psychosis and schizophrenia.

Chemical Identifiers

CAS 1432043-31-6
Molecular Formula C32H37Cl2N7O4

Specifications

Quantity 10 mg
Product Line Tocris
Formula Weight 654.59
Purity >97%
Molecular Weight (g/mol) 654.593
Chemical Name or Material WAY 267464 dihydrochloride
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.