accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 143520-19-8

IUPAC Name: [(dimethylamino)({[3,4,5-tris(benzyloxy)-6-[(benzyloxy)methyl]oxan-2-yl]oxy})phosphoryl]dimethylamine
Synonyms: [dimethylamino([3,4,5-tris(benzyloxy)-6-[(benzyloxy)methyl]oxan-2-yl]oxy)phosphoryl]dimethylamine
Molecular Weight (g/mol): 674.78
Molecular Formula: C38H47N2O7P
InChi Key: RTYAYXUZHPWJOK-UHFFFAOYNA-N
SMILES: CN(C)P(=O)(OC1OC(COCC2=CC=CC=C2)C(OCC2=CC=CC=C2)C(OCC2=CC=CC=C2)C1OCC1=CC=CC=C1)N(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results