CAS RN 1435467-38-1
IUPAC Name:
2-[6-(2,5-dimethoxyphenyl)-4-oxo-2-sulfanylidene-1,2,3,4-tetrahydropyrimidin-1-yl]acetamide
Synonyms:
2-[6-(2,5-dimethoxyphenyl)-4-oxo-2-sulfanylidene-3H-pyrimidin-1-yl]acetamide
Molecular Weight (g/mol):
321.35
Molecular Formula:
C14H15N3O4S
InChi Key:
LJBUZOGABRDGBR-UHFFFAOYSA-N
SMILES:
COC1=CC(=C(OC)C=C1)C1=CC(=O)NC(=S)N1CC(N)=O
1
–
1
of
1
results
1
Promotions Available
Tocris Bioscience™ PF 06281355, >98%
Tocris Bioscience™ PF 06281355, >98%
CAS: 1435467-38-1 Molecular Formula: C14H15N3O4S