CAS RN 143556-24-5
IUPAC Name:
8-amino-5-chloro-7-phenyl-1H,2H,3H,4H-pyrido[3,4-d]pyridazine-1,4-dione sodium
Synonyms:
8-amino-5-chloro-7-phenyl-2H,3H-pyrido[3,4-d]pyridazine-1,4-dione sodium
Molecular Weight (g/mol):
311.68
Molecular Formula:
C13H9ClN4NaO2
InChi Key:
WRVLDBOUZVBBCX-UHFFFAOYSA-N
SMILES:
[Na].NC1=C2C(=O)NNC(=O)C2=C(Cl)N=C1C1=CC=CC=C1
1
–
1
of
1
results
1
R&D Systems™ L 012 sodium salt
CAS: 143556-24-5 Molecular Formula: C13H9ClN4NaO2