CAS RN 143692-48-2
IUPAC Name:
hydrogen 10-(4-aminophenyl)-N,13-dimethyl-4,6-dioxa-11,12-diazatricyclo[7.5.0.0³,⁷]tetradeca-1,3(7),8,10-tetraene-12-carboxamide chloride
Synonyms:
hydrogen 10-(4-aminophenyl)-N,13-dimethyl-4,6-dioxa-11,12-diazatricyclo[7.5.0.0³,⁷]tetradeca-1,3(7),8,10-tetraene-12-carboxamide chloride
Molecular Weight (g/mol):
388.85
Molecular Formula:
C19H21ClN4O3
InChi Key:
ASLCSBBDVWPSQT-UHFFFAOYNA-N
SMILES:
[H+].[Cl-].CNC(=O)N1N=C(C2=CC=C(N)C=C2)C2=CC3=C(OCO3)C=C2CC1C
1
–
1
of
1
results
1
R&D Systems™ GYKI 53655 hydrochloride
CAS: 143692-48-2 Molecular Formula: C19H21ClN4O3