accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SO

CAS RN 1438-91-1

IUPAC Name: 2-[(methylsulfanyl)methyl]furan
Synonyms: 2-[(methylsulfanyl)methyl]furan
Molecular Weight (g/mol): 128.19
Molecular Formula: C6H8OS
InChi Key: SKSFHXVDHVKIBN-UHFFFAOYSA-N
SMILES: CSCC1=CC=CO1
Boiling Point 
  • (3)
  • (1)
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)