CAS RN 143851-98-3
IUPAC Name:
hydrogen N-{4-[2-(6,7-dimethoxy-1,2,3,4-tetrahydroisoquinolin-2-yl)ethyl]phenyl}-5-methoxy-9-oxo-9,10-dihydroacridine-4-carboxamide chloride
Synonyms:
hydrogen N-{4-[2-(6,7-dimethoxy-3,4-dihydro-1H-isoquinolin-2-yl)ethyl]phenyl}-5-methoxy-9-oxo-10H-acridine-4-carboxamide chloride
Molecular Weight (g/mol):
600.11
Molecular Formula:
C34H34ClN3O5
InChi Key:
IQOJZZHRYSSFJM-UHFFFAOYSA-N
SMILES:
[H+].[Cl-].COC1=C2NC3=C(C=CC=C3C(=O)C2=CC=C1)C(=O)NC1=CC=C(CCN2CCC3=CC(OC)=C(OC)C=C3C2)C=C1
1
–
1
of
1
results
1
R&D Systems™ Elacridar hydrochloride
CAS: 143851-98-3 Molecular Formula: C34H34ClN3O5