CAS RN 1440251-53-5
CAS RN 1440251-53-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Tocris Bioscience™ ML 315 hydrochloride
Low molecular weight, organic compound that may help regulate biological process.