Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ ML 315 hydrochloride
GREENER_CHOICESDP

Catalog No. 570050 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
57-005-0 50 mg
57-001-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 57-005-0 Supplier Tocris Bioscience™ Supplier No. 5700/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Low molecular weight, organic compound that may help regulate biological process.

ML 315 hydrochloride is a inhibitor of cdc2-like kinases (Clk) and DYRK kinases (IC50 values are 68, 68 and 231 nM for Clk1, Clk4 and Clk2, respectively, and 282 and 1156 nM for Dyrk1A and Dyrk1B). Shows 20-fold selectivity for Clk4 over Dyrk1A. Stable in both mouse and human plasma. Moderately cell permeable.

Specifications

CAS 1440251-53-5
Quantity 50 mg
Target Cdc2-like Kinase
Molecular Formula C18H13Cl2N3O2.HCl
Formula Weight 410.68g/mol
Product Type Inhibitor

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.