accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2NNN(R)O(R)OH(S)HO(R)HO

CAS RN 14432-09-8

IUPAC Name: (2R,3R,4S,5R)-2-{7-amino-3H-imidazo[4,5-b]pyridin-3-yl}-5-(hydroxymethyl)oxolane-3,4-diol
Synonyms: (2R,3R,4S,5R)-2-{7-aminoimidazo[4,5-b]pyridin-3-yl}-5-(hydroxymethyl)oxolane-3,4-diol
Molecular Weight (g/mol): 266.26
Molecular Formula: C11H14N4O4
InChi Key: NVUDDRWKCUAERS-PNHWDRBUSA-N
SMILES: NC1=C2N=CN([C@@H]3O[C@H](CO)[C@@H](O)[C@H]3O)C2=NC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)