Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ 1-Deazaadenosine
GREENER_CHOICESDP

Catalog No. 448810 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
44-881-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 44-881-0 Supplier R&D Systems™ Supplier No. 4488/10

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Adenosine deaminase inhibitor

Inhibitor of adenosine deaminase (Ki = 0.66 μM). Demonstrates antitumor activity in a range of leukemia cell lines.

Chemical Identifiers

CAS 14432-09-8
Molecular Formula C11H14N4O4

Specifications

Quantity 10 mg
Product Line Tocris
Formula Weight 266.25
Purity >98%
Molecular Weight (g/mol) 266.257
Chemical Name or Material 1-Deazaadenosine
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.