CAS RN 1448858-83-0
IUPAC Name:
[({[3,4-dihydroxy-5-(5-iodo-2,4-dioxo-1,2,3,4-tetrahydropyrimidin-1-yl)oxolan-2-yl]methoxy}(hydroxy)phosphoryl)oxy]phosphonic acid trisodium
Synonyms:
{[3,4-dihydroxy-5-(5-iodo-2,4-dioxo-3H-pyrimidin-1-yl)oxolan-2-yl]methoxy(hydroxy)phosphoryl}oxyphosphonic acid trisodium
Molecular Weight (g/mol):
599.03
Molecular Formula:
C9H13IN2Na3O12P2
InChi Key:
WAJPEYKEXKCDGT-UHFFFAOYNA-N
SMILES:
[Na].[Na].[Na].OC1C(O)C(OC1COP(O)(=O)OP(O)(O)=O)N1C=C(I)C(=O)NC1=O
1
–
1
of
1
results
1
R&D Systems™ MRS 2693 trisodium salt
CAS: 1448858-83-0 Molecular Formula: C9H13IN2Na3O12P2