accessibility menu, dialog, popup

UserName

Viewing profile as user:
SNSSNS

CAS RN 1450644-28-6

IUPAC Name: 5-(1,2-thiazol-5-yldisulfanyl)-1,2-thiazole
Synonyms: 5-(1,2-thiazol-5-yldisulfanyl)-1,2-thiazole
Molecular Weight (g/mol): 232.35
Molecular Formula: C6H4N2S4
InChi Key: MLRAMCAHIWHWRM-UHFFFAOYSA-N
SMILES: S(SC1=CC=NS1)C1=CC=NS1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)