Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ NU 9056
GREENER_CHOICESDP

Catalog No. 490310 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
49-031-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 49-031-0 Supplier R&D Systems™ Supplier No. 4903/10

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Inhibitor of KAT5 (Tip60)

Selective KAT5 (Tip60) histone acetyltransferase inhibitor (IC50 values are < 2, 60, 36, and >100 μM for KAT5, p300, pCAF and GCN5, respectively). Inhibits protein acetylation in prostate cancer cell lines and blocks DNA damage response. Decreases proliferation of LNCaP cells; induces apoptosis via caspase activation.

Chemical Identifiers

CAS 1450644-28-6
Molecular Formula C6H4N2S4

Specifications

Quantity 10 mg
Product Line Tocris
Formula Weight 232.37
Purity >98%
Molecular Weight (g/mol) 232.35
Chemical Name or Material NU 9056
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.