CAS RN 145148-39-6
IUPAC Name:
1-[(2-methoxyphenyl)methyl]-2-phenylpiperidin-3-amine dihydrochloride
Synonyms:
1-[(2-methoxyphenyl)methyl]-2-phenylpiperidin-3-amine dihydrochloride
Molecular Weight (g/mol):
369.33
Molecular Formula:
C19H26Cl2N2O
InChi Key:
STUCSMJJXNCIOE-UHFFFAOYNA-N
SMILES:
Cl.Cl.COC1=CC=CC=C1CN1CCCC(N)C1C1=CC=CC=C1
1
–
1
of
1
results
1
R&D Systems™ CP 99994 dihydrochloride
CAS: 145148-39-6 Molecular Formula: C19H26Cl2N2O