accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OONOHCl

CAS RN 1454-53-1

IUPAC Name: ethyl 1-benzyl-4-oxopiperidine-3-carboxylate hydrochloride
Synonyms: ethyl 1-benzyl-4-oxopiperidine-3-carboxylate hydrochloride
Molecular Weight (g/mol): 297.78
Molecular Formula: C15H20ClNO3
InChi Key: YPFMNHZRNXPYBG-UHFFFAOYNA-N
SMILES: Cl.CCOC(=O)C1CN(CC2=CC=CC=C2)CCC1=O
Percent Purity 
  • (1)
  • (2)
Physical Form 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)