accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1454889-37-2

IUPAC Name: ({5-[6-(methylamino)-9H-purin-9-yl]-2-[(phosphonooxy)methyl]oxolan-3-yl}oxy)phosphonic acid tetrasodium
Synonyms: {5-[6-(methylamino)purin-9-yl]-2-[(phosphonooxy)methyl]oxolan-3-yl}oxyphosphonic acid tetrasodium
Molecular Weight (g/mol): 517.19
Molecular Formula: C11H17N5Na4O9P2
InChi Key: HLJPLHFSEAJJGC-UHFFFAOYNA-N
SMILES: [Na].[Na].[Na].[Na].CNC1=C2N=CN(C3CC(OP(O)(O)=O)C(COP(O)(O)=O)O3)C2=NC=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results